Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-(Azodicarbonyl)dipiperidine 97%

Supplier: Origen Biomedical, Inc
Warning

H27719-14 H27719-06
AAH27719-14EA 308 USD InStock
AAH27719-14 AAH27719-06
1,1'-(Azodicarbonyl)dipiperidine 97%
1,1'-(Azodicarbonyl)dipiperidine
Purity: 97% CAS No.: 10465-81-3 Molecular Formula: C12H20N4O2 Formula Weight: 252.32 Odor: Odorless Melting Point: 133-136° MDL No.: MFCD00010111


Formula: C₁₂H₂₀N₄O₂
MW: 252.32 g/mol
Melting Pt: 133…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00010111
CAS Number: 10465-81-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty