H27748-03
H27748-06
AAH27748-03EA
74
USD
InStock
AAH27748-03
AAH27748-06
2-Chloropyridine-6-carbonitrile 97%
2-Chloropyridine-6-carbonitrile
Purity: 97%
CAS No.: 33252-29-8
Molecular Formula: C6H3ClN2
Formula Weight: 138.56
Melting Point: 85-88°
UN No.: 3439
MDL No.: MFCD00274527