H28343-03
H28343-06
AAH28343-03EA
61.16
USD
InStock
AAH28343-03
AAH28343-06
5-Hydroxy-1,4-naphthoquinone (juglone) 99%
5-Hydroxy-1,4-naphthoquinone (juglone)
Purity: 99%
CAS No.: 481-39-0
Molecular Formula: C10H6O3
Formula Weight: 174.16
Melting Point: 161-163°
Merck Reference: 14,5629
MDL No.: MFCD00001684