H28374-03
H28374-MD
AAH28374-03EA
425
USD
InStock
AAH28374-03
AAH28374-MD
AAH28374-MC
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 97%
(R,R)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Purity: 97%
CAS No.: 55739-58-7
Molecular Formula: C28H28O2P2
Formula Weight: 458.47
Melting Point: 101-105°
Optical Rotation: -83° (c=1 in chloroform)
MDL No.: MFCD05863546