Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Pyrimidineboronic acid hemihydrate 97%

Supplier: Origen Biomedical, Inc
Warning

H29212-03 H29212-09
AAH29212-03EA 106 USD InStock
AAH29212-03 AAH29212-09
5-Pyrimidineboronic acid hemihydrate 97%
5-Pyrimidineboronic acid hemihydrate
Purity: 97% CAS No.: 696602-91-2 Molecular Formula: C4H5BN2O2.0.5H2O Formula Weight: 132.91 (123.91anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° MDL No.: MFCD03002366


Formula: C₄H₅BN₂O₂·0.5H₂O
MW: 132.91 g/mol (123.91 anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03002366
CAS Number: 696602-91-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty