Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Thiocarbonyldi-2(1H)-pyridone 95%

Supplier: Origen Biomedical, Inc
Warning

H30087-03 H30087-06
AAH30087-03EA 17.7 USD InStock
AAH30087-03 AAH30087-06
1,1'-Thiocarbonyldi-2(1H)-pyridone 95%
1,1'-Thiocarbonyldi-2(1H)-pyridone
Purity: 95% CAS No.: 102368-13-8 Molecular Formula: C11H8N2O2S Formula Weight: 232.26 Melting Point: 163-166° MDL No.: MFCD00075238


Formula: C₁₁H₈N₂O₂S
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00075238
CAS Number: 102368-13-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty