Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromo-4-hydroxybenzoate 98%

Supplier: Origen Biomedical, Inc
Warning

H30222-09 H30222-18
AAH30222-09EA 57.8 USD InStock
AAH30222-09 AAH30222-18
Methyl-3-bromo-4-hydroxybenzoate 98%
Methyl-3-bromo-4-hydroxybenzoate
Purity: 98% CAS No.: 29415-97-2 Molecular Formula: C8H7BrO3 Formula Weight: 231.05 Melting Point: 108-110° MDL No.: MFCD06203850


Formula: C₈H₇BrO₃
MW: 231.05
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD06203850
CAS Number: 29415-97-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty