Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methylphenylboronic acid 97%

Supplier: Origen Biomedical, Inc
Warning

H30893-03 H30893-MD
AAH30893-03EA 172 USD InStock
AAH30893-03 AAH30893-MD
3-Chloro-2-methylphenylboronic acid 97%
3-Chloro-2-methylphenylboronic acid
Purity: 97% CAS No.: 313545-20-9 Molecular Formula: C7H8BClO2 Formula Weight: 170.40 MDL No.: MFCD04115641


Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115641
CAS Number: 313545-20-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty