Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminobenzo[d]thiazole-6-carboxylic acid 95%

Supplier: Origen Biomedical, Inc
Warning

H31512-03 H31512-06
AAH31512-03EA 87.3 USD InStock
AAH31512-03 AAH31512-06
2-Aminobenzo[d]thiazole-6-carboxylic acid 95%
2-Aminobenzo[d]thiazole-6-carboxylic acid
Purity: 95% CAS No.: 93-85-6 Molecular Formula: C8H6N2O2S Formula Weight: 194.21 Melting Point: 300° MDL No.: MFCD00054180


Formula: C₈H₆N₂O₂S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00054180
CAS Number: 93-85-6
EINECS: 202-283-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty