Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxalyl dichloride 2 M in dichloromethane

Supplier: Origen Biomedical, Inc
Danger

H31617-AE H31617-AP
AAH31617-AEEA 107.57 USD InStock
AAH31617-AE AAH31617-AP
Oxalyl dichloride 2 M in dichloromethane
Oxalyl dichloride
CAS No.: 79-37-8 Molecular Formula: C2Cl2O2 Formula Weight: 126.93 UN No.: 2922 MDL No.: MFCD00000704


Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63
Melting Pt: -12
Density: 1.335
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8 III
Merck Index: 12,07046
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty