Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-2-(trifluoromethyl)benzonitrile 98%

Supplier: Origen Biomedical, Inc
Danger

H31904-03 H31904-06
AAH31904-03EA 80.2 USD InStock
AAH31904-03 AAH31904-06
4-Methyl-2-(trifluoromethyl)benzonitrile 98%
4-Methyl-2-(trifluoromethyl)benzonitrile
Purity: 98% CAS No.: 261952-05-0 Molecular Formula: C9H6F3N Formula Weight: 185.15 Melting Point: 22-28° UN No.: 3439 MDL No.: MFCD01631596


Formula: C₉H₆F₃N
MW: 185.15
Melting Pt: 22…28 °C
Storage Temperature: Ambient
MDL Number: MFCD01631596
CAS Number: 261952-05-0
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty