Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-methoxybenzoic acid 98%

Supplier: Origen Biomedical, Inc
Warning

H31960-03 H31960-06
AAH31960-03EA 22.2 USD InStock
AAH31960-03 AAH31960-06
3-Chloro-5-methoxybenzoic acid 98%
3-Chloro-5-methoxybenzoic acid
Purity: 98% CAS No.: 82477-67-6 Molecular Formula: C8H7ClO3 Formula Weight: 186.59 MDL No.: MFCD01317994


Formula: C₈H₇ClO₃
MW: 186.59
Storage Temperature: Ambient
MDL Number: MFCD01317994
CAS Number: 82477-67-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty