H32013-18
H32013-09
AAH32013-18EA
163
USD
InStock
AAH32013-18
AAH32013-09
2-Furaldehyde-2,2-dimethylhydrazone 98%
2-Furaldehyde-2,2-dimethylhydrazone
Purity: 98%
CAS No.: 14064-21-2
Molecular Formula: C7H10N2O
Formula Weight: 138.17
Flash Point: 92°(198°F)
Refractive Index: 1.5800
Odor: Recognizable
Boiling Point: 69°/2mm
MDL No.: MFCD00010685