Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Trimethylthioacetamide 97%

Supplier: Origen Biomedical, Inc

H32092-09 H32092-03
AAH32092-09EA 346 USD InStock
AAH32092-09 AAH32092-03
2,2,2-Trimethylthioacetamide 97%
2,2,2-Trimethylthioacetamide
Purity: 97% CAS No.: 630-22-8 Molecular Formula: C5H11NS Formula Weight: 117.21 Melting Point: 113-117° MDL No.: MFCD09742834


Formula: C₅H₁₁NS
MDL Number: MFCD09742834
CAS Number: 630-22-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty