Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromaminic acid ≥90%

Supplier: Origen Biomedical, Inc
Warning

H32110-14 H32110-22
AAH32110-14EA 37.8 USD InStock
AAH32110-14 AAH32110-22
Bromaminic acid ≥90%
Bromaminic acid
Purity: 90+% CAS No.: 116-81-4 Molecular Formula: C14H8BrNO5S Formula Weight: 382.19 Odor: Odorless Melting Point: ca 280° dec. MDL No.: MFCD00035694


Formula: C₁₄H₈BrNO₅S
Melting Pt: ca 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035694
CAS Number: 116-81-4
EINECS: 204-159-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty