Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (4-fluorophenyl)acetate 98%

Supplier: Origen Biomedical, Inc
Warning

H32236-06 H32236-14
AAH32236-06EA 53.3 USD InStock
AAH32236-06 AAH32236-14
Methyl (4-fluorophenyl)acetate 98%
Methyl (4-fluorophenyl)acetate
Purity: 98% CAS No.: 34837-84-8 Molecular Formula: C9H9FO2 Formula Weight: 168.17 MDL No.: MFCD00800608


Formula: C₉H₉FO₂
MW: 168.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800608
CAS Number: 34837-84-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty