Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Vinylbenzoic acid 96%

Supplier: Origen Biomedical, Inc
Warning

H32556-03 H32556-06
AAH32556-03EA 66.7 USD InStock
AAH32556-03 AAH32556-06
3-Vinylbenzoic acid 96%
3-Vinylbenzoic acid
Purity: 96% CAS No.: 28447-20-3 Molecular Formula: C9H8O2 Formula Weight: 148.16 Storage Temperature: -30°C to -10°C Melting Point: 95° MDL No.: MFCD00157038


Formula: C₉H₈O₂
MW: 148.16 g/mol
Melting Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00157038
CAS Number: 28447-20-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty