H32610-03
H32610-09
AAH32610-03EA
130
USD
InStock
AAH32610-03
AAH32610-09
4-Fluoro-2-nitrobenzonitrile 97%
4-Fluoro-2-nitrobenzonitrile
Purity: 97%
CAS No.: 80517-21-1
Molecular Formula: C7H3FN2O2
Formula Weight: 166.11
Melting Point: 69-73°
UN No.: 3439
MDL No.: MFCD00277447