Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodo-1-phenyl-1H-pyrazole 95%

Supplier: Origen Biomedical, Inc
Warning

H32612-03 H32612-MD
AAH32612-03EA 212 USD InStock
AAH32612-03 AAH32612-MD
4-Iodo-1-phenyl-1H-pyrazole 95%
4-Iodo-1-phenyl-1H-pyrazole
Purity: 95% CAS No.: 23889-85-2 Molecular Formula: C9H7IN2 Formula Weight: 270.07 MDL No.: MFCD11707168


Formula: C₉H₇IN₂
Storage Temperature: Ambient
MDL Number: MFCD11707168
CAS Number: 23889-85-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty