Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentanesulfonyl chloride 97%

Supplier: Origen Biomedical, Inc
Danger

H32865-03 H32865-MD H32865-06
AAH32865-03EA 180 USD InStock
AAH32865-03 AAH32865-MD AAH32865-06
Cyclopentanesulfonyl chloride 97%
Cyclopentanesulphonyl chloride
Purity: 97% CAS No.: 26394-17-2 Molecular Formula: C5H9ClO2S Formula Weight: 168.64 Boiling Point: 71-73°/2mm UN No.: 3265 MDL No.: MFCD04117774


Formula: C₅H₉ClO₂S
Boiling Pt: 71…73 °C/2 mm
Storage Temperature: Ambient
MDL Number: MFCD04117774
CAS Number: 26394-17-2
UN: 3265
ADR: "8",III
Order Now
Specification Test Results
Assay 97%

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty