Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2,3-difluorotoluene 95%

Supplier: Origen Biomedical, Inc
Warning

H32941-03 H32941-06
AAH32941-03EA 95 USD InStock
AAH32941-03 AAH32941-06
6-Bromo-2,3-difluorotoluene 95%
6-Bromo-2,3-difluorotoluene
Purity: 97% CAS No.: 847502-81-2 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Refractive Index: 1.5105


Formula: C₇H₅BrF₂
Storage Temperature: Ambient
MDL Number: MFCD07777161
CAS Number: 847502-81-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty