Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate 96%

Supplier: Origen Biomedical, Inc
Warning

H33196-06 H33196-14
AAH33196-06EA 98.8 USD InStock
AAH33196-06 AAH33196-14
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate 96%
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Purity: 96% CAS No.: 4815-29-6 Molecular Formula: C10H13NO2S Formula Weight: 211.28 Melting Point: 95-96° MDL No.: MFCD00102063


Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00102063
CAS Number: 4815-29-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty