Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2,6-Dichlorophenyl)propionic acid 96%

Supplier: Origen Biomedical, Inc
Warning

H33210-03 H33210-06 H33210-14
AAH33210-03EA 16.1 USD InStock
AAH33210-03 AAH33210-06 AAH33210-14
3-(2,6-Dichlorophenyl)propionic acid 96%
3-(2,6-Dichlorophenyl)propionic acid
Purity: 96% CAS No.: 51656-68-9 Molecular Formula: C9H8Cl2O2 Formula Weight: 219.07 Melting Point: 97-99° MDL No.: MFCD01310807


Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01310807
CAS Number: 51656-68-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty