Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-2-thienyl sulfide 97%

Supplier: Origen Biomedical, Inc

H33284-03 H33284-06
AAH33284-03EA 39.9 USD InStock
AAH33284-03 AAH33284-06
Di-2-thienyl sulfide 97%
Di-2-thienyl sulfide
Purity: 97% CAS No.: 3988-99-6 Molecular Formula: C8H6S3 Formula Weight: 198.32 Flash Point: 100°(212°F) Refractive Index: 1.6490 Boiling Point: 102-104°/0.9mm MDL No.: MFCD00052047


Formula: C₈H₆S₃
Boiling Pt: 102…104 °C/0.9 mm
Density: 1.311
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052047
CAS Number: 3988-99-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty