Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminopyridine-6-methanol 97%

Supplier: Origen Biomedical, Inc
Warning

H33485-03 H33485-06
AAH33485-03EA 200 USD InStock
AAH33485-03 AAH33485-06 AAH33485-MD
2-Aminopyridine-6-methanol 97%
2-Aminopyridine-6-methanol
Purity: 97% CAS No.: 79651-64-2 Molecular Formula: C6H8N2O Formula Weight: 124.14 Melting Point: 87-92° MDL No.: MFCD09863097


Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 87…92 °C
Storage Temperature: Ambient
MDL Number: MFCD09863097
CAS Number: 79651-64-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty