H33546-03
H33546-09
AAH33546-03EA
16.7
USD
InStock
AAH33546-03
AAH33546-09
4-Nitrophthalonitrile 97%
4-Nitrophthalonitrile
Purity: 97%
CAS No.: 31643-49-9
Molecular Formula: C8H3N3O2
Formula Weight: 173.13
Melting Point: 136-137°
MDL No.: MFCD00040301