Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Thiophen-2-yl)benzoic acid 96%

Supplier: Origen Biomedical, Inc
Warning

H33642-03 H33642-06
AAH33642-03EA 70 USD InStock
AAH33642-03 AAH33642-06
4-(Thiophen-2-yl)benzoic acid 96%
4-(Thiophen-2-yl)benzoic acid
Purity: 96% CAS No.: 29886-62-2 Molecular Formula: C11H8O2S Formula Weight: 204.25 Melting Point: 243-245° MDL No.: MFCD00690742


Formula: C₁₁H₈O₂S
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00690742
CAS Number: 29886-62-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty