Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 96%

Supplier: Origen Biomedical, Inc

H34131-03 H34131-06
AAH34131-03EA 47 USD InStock
AAH34131-03 AAH34131-06 AAH34131-MD
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 96%
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one
Purity: 96% CAS No.: 179688-01-8 Molecular Formula: C16H16N2O3 Formula Weight: 284.31 MDL No.: MFCD04115119


Formula: C₁₆H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04115119
CAS Number: 179688-01-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty