Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-2-fluoroacetophenone 98%

Supplier: Origen Biomedical, Inc
Danger

H34179-03 H34179-09
AAH34179-03EA 36.6 USD InStock
AAH34179-03 AAH34179-09
ω-Bromo-2-fluoroacetophenone 98%
ω-Bromo-2-fluoroacetophenone
Purity: 98% CAS No.: 655-15-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 UN No.: 3265 MDL No.: MFCD00278796


Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD00278796
CAS Number: 655-15-2
UN: 3265
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty