Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxypicolinic acid 97%

Supplier: Origen Biomedical, Inc
Warning

H34262-03 H34262-06 H34262-MD
AAH34262-03EA 178 USD InStock
AAH34262-03 AAH34262-06 AAH34262-MD
4-Hydroxypicolinic acid 97%
4-Hydroxypicolinic acid
Purity: 97% CAS No.: 22468-26-4 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Melting Point: ca 280° dec. MDL No.: MFCD00521155


Formula: C₆H₅NO₃
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00521155
CAS Number: 22468-26-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty