Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Thienyl)benzenesulfonyl chloride 96%

Supplier: Origen Biomedical, Inc
Danger

H34399-03 H34399-06 H34399-MD
AAH34399-03EA 106 USD InStock
AAH34399-03 AAH34399-06 AAH34399-MD
4-(2-Thienyl)benzenesulfonyl chloride 96%
4-(2-Thienyl)benzenesulfonyl chloride
Purity: 96% CAS No.: 209592-91-6 Molecular Formula: C10H7ClO2S2 Formula Weight: 258.75 Melting Point: 107-108° UN No.: 3261 MDL No.: MFCD11655569


Formula: C₁₀H₇ClO₂S₂
MW: 258.75 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD11655569
CAS Number: 209592-91-6
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty