H34492-03
H34492-06
AAH34492-03EA
40.5
USD
InStock
AAH34492-03
AAH34492-06
3-(2-Methoxyphenyl)-1H-pyrazole 97%
3-(2-Methoxyphenyl)-1H-pyrazole
Purity: 97%
CAS No.: 59843-63-9
Molecular Formula: C10H10N2O
Formula Weight: 174.20
Melting Point: 82-84°
MDL No.: MFCD02091524