Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine 97%

Supplier: Origen Biomedical, Inc
Warning

H50873-03 H50873-MD
AAH50873-03EA 189 USD InStock
AAH50873-03 AAH50873-MD
4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine 97%
4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine
Purity: 97% CAS No.: 733748-92-0 Molecular Formula: C10H17N3O Formula Weight: 195.27 Refractive Index: 1.4860 MDL No.: MFCD08061038


Formula: C₁₀H₁₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD08061038
CAS Number: 733748-92-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty