H51050-03
H51050-06
AAH51050-03EA
115
USD
InStock
AAH51050-03
AAH51050-06
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine
Purity: 96%
CAS No.: 61478-26-0
Molecular Formula: C10H19NO4
Formula Weight: 217.26
MDL No.: MFCD02094386