Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hydroxymethyl)phenylboronic acid pinacol ester

Supplier: Origen Biomedical, Inc

H51126-06 H51126-14 H51126-03
AAH51126-06EA 115 USD InStock
AAH51126-06 AAH51126-14 AAH51126-03
3-(Hydroxymethyl)phenylboronic acid pinacol ester
3-(Hydroxymethyl)phenylboronic acid pinacol ester
CAS No.: 443776-76-9 Molecular Formula: C13H19BO3 Formula Weight: 234.10 MDL No.: MFCD09266196


Formula: C₁₃H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD09266196
CAS Number: 443776-76-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty