H51133-06
H51133-14
AAH51133-06EA
32.8
USD
InStock
AAH51133-06
AAH51133-14
N,N'-Dimorpholinomethane 98%
N,N'-Dimorpholinomethane
Purity: 98%
CAS No.: 5625-90-1
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
Boiling Point: 122-124°/12mm
MDL No.: MFCD00023369