Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Propylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Origen Biomedical, Inc
Danger

H51162-AE H51162-AP
AAH51162-AEEA 71.98 USD InStock
AAH51162-AE AAH51162-AP
n-Propylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
n-Propylmagnesium chloride
CAS No.: 2234-82-4 Molecular Formula: CH3CH2CH2MgCl Formula Weight: 102.85 UN No.: 3399 MDL No.: MFCD00000474


Formula: C₃H₇ClMg
Storage Temperature: Ambient
MDL Number: MFCD00000474
CAS Number: 2234-82-4
UN: 3399
ADR: 4.3,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty