H51258-03
H51258-06
AAH51258-03EA
98.3
USD
InStock
AAH51258-03
AAH51258-06
cis-1-Ethoxy-2-(tributylstannyl)ethene 94%
cis-1-Ethoxy-2-(tributylstannyl)ethene
Purity: 94%
CAS No.: 64724-29-4
Molecular Formula: C16H34OSn
Formula Weight: 361.15
Melting Point: 84-87°
UN No.: 2788
MDL No.: MFCD07787391