Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Tributylstannyl)pyridine 96%

Supplier: Origen Biomedical, Inc
Danger

H51435-03 H51435-06
AAH51435-03EA 83.5 USD InStock
AAH51435-03 AAH51435-06
4-(Tributylstannyl)pyridine 96%
4-(Tributylstannyl)pyridine
Purity: 96% CAS No.: 124252-41-1 Molecular Formula: C17H31NSn Formula Weight: 368.15 UN No.: 2788 MDL No.: MFCD01319071


Formula: C₁₇H₃₁NSn
Storage Temperature: Ambient
MDL Number: MFCD01319071
CAS Number: 124252-41-1
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty