Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride

Supplier: Origen Biomedical, Inc
Warning

H51664-03 H51664-06 H51664-MD
AAH51664-03EA 144 USD InStock
AAH51664-03 AAH51664-06 AAH51664-MD
3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride
3-Methyl-5-(3-pyrrolidinyl)-1,2,4-oxadiazole hydrochloride
CAS No.: 1121057-52-0 Molecular Formula: C7H11N3O.HCl Formula Weight: 189.65 MDL No.: MFCD11870728


Formula: C₇H₁₁N₃O·HCl
Storage Temperature: Ambient
MDL Number: MFCD11870728
CAS Number: 1121057-52-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty