Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl thioacetimidate hydrochloride 96%

Supplier: Origen Biomedical, Inc
Danger

H51844-03 H51844-06 H51844-14
AAH51844-03EA 27.4 USD InStock
AAH51844-03 AAH51844-06 AAH51844-14
Benzyl thioacetimidate hydrochloride 96%
Benzyl thioacetimidate hydrochloride


Formula: C₉H₁₁NS·HCl
Storage Temperature: Ambient
MDL Number: MFCD05865210
CAS Number: 32894-07-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty