Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(N,N-Diphenylamino)phenylboronic acid pinacol ester 95%

Supplier: Origen Biomedical, Inc

H51946-03 H51946-06
AAH51946-03EA 59.3 USD InStock
AAH51946-03 AAH51946-06
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester 95%
4-(N,N-Diphenylamino)phenylboronic acid pinacol ester
Purity: 95% Molecular Formula: C24H26BNO2 Formula Weight: 371.29 MDL No.: MFCD13195770


Formula: C₂₄H₂₆BNO₂
MW: 371.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195770
CAS Number: 267221-88-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty