Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(N-beta-Boc)-L-alpha,beta-diaminopropionic acid 95%

Supplier: Origen Biomedical, Inc

H51987-03 H51987-06 H51987-MD
AAH51987-03EA 119 USD InStock
AAH51987-03 AAH51987-06 AAH51987-MD
Fmoc-(N-beta-Boc)-L-alpha,beta-diaminopropionic acid 95%
Fmoc-Dap(Boc)-OH
Purity: 95% CAS No.: 162558-25-0 Molecular Formula: C23H26N2O6 Formula Weight: 426.47 Storage Temperature: 2°C to 8°C Beilstein Reference: 7057792 MDL No.: MFCD00235896


Formula: C₂₃H₂₆N₂O₆
MW: 426.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235896
CAS Number: 162558-25-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty