Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%

Supplier: Origen Biomedical, Inc

H51988-03 H51988-06 H51988-MD
AAH51988-03EA 89.7 USD InStock
AAH51988-03 AAH51988-06 AAH51988-MD
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 95%
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine
Purity: 95% CAS No.: 114873-01-7 Molecular Formula: C14H18FNO4 Formula Weight: 283.30 Melting Point: 75-80° Beilstein Reference: 5347375 Optical Rotation: +6.5° (c=1 in methanol) MDL No.: MFCD00672522


Formula: C₁₄H₁₈FNO₄
Melting Pt: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00672522
CAS Number: 114873-01-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty