Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,3R)-3-Aminocyclopentanecarboxylic acid 95%

Supplier: Origen Biomedical, Inc
Warning

H52040-03 H52040-MD
AAH52040-03EA 333 USD InStock
AAH52040-03 AAH52040-MD
(1S,3R)-3-Aminocyclopentanecarboxylic acid 95%
(1S,3R)-3-Aminocyclopentanecarboxylic acid
Purity: 95% CAS No.: 71830-07-4 Molecular Formula: C6H11NO2 Formula Weight: 129.16 MDL No.: MFCD00211290


Formula: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00211290
CAS Number: 71830-07-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty