Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fmoc-3-(4-thiazolyl)-L-alanine 95%

Supplier: Origen Biomedical, Inc

H52071-03 H52071-06 H52071-MD
AAH52071-03EA 194 USD InStock
AAH52071-03 AAH52071-06 AAH52071-MD
N-Fmoc-3-(4-thiazolyl)-L-alanine 95%
N-Fmoc-3-(4-thiazolyl)-L-alanine
Purity: 95% CAS No.: 205528-32-1 Molecular Formula: C21H18N2O4S Formula Weight: 394.44 Melting Point: 178-179° MDL No.: MFCD00672568


Formula: C₂₁H₁₈N₂O₄S
Melting Pt: 178…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00672568
CAS Number: 205528-32-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty