Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-O-(benzylphospho)-L-serine 95%

Supplier: Origen Biomedical, Inc

H52168-03 H52168-06 H52168-MD
AAH52168-03EA 232 USD InStock
AAH52168-03 AAH52168-06 AAH52168-MD
Fmoc-O-(benzylphospho)-L-serine 95%
Fmoc-O-(benzylphospho)-L-serine
Purity: 95% CAS No.: 158171-14-3 Molecular Formula: C25H24NO8P Formula Weight: 497.44 Beilstein Reference: 7060483 MDL No.: MFCD00797869


Formula: C₂₅H₂₄NO₈P
MW: 497.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797869
CAS Number: 158171-14-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty