Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-L-beta-homophenylalanine 95%

Supplier: Origen Biomedical, Inc

H52174-03 H52174-MD
AAH52174-03EA 276 USD InStock
AAH52174-03 AAH52174-MD
Boc-L-beta-homophenylalanine 95%
Boc-L-beta-homophenylalanine
Purity: 95% CAS No.: 51871-62-6 Molecular Formula: C15H21NO4 Formula Weight: 279.34 Melting Point: 101-107° Beilstein Reference: 3060457 Optical Rotation: -19.0° (c=1 in dichloromethane) MDL No.: MFCD01076271


Formula: C₁₅H₂₁NO₄
Melting Pt: 101…107 °C
Storage Temperature: Ambient
MDL Number: MFCD01076271
CAS Number: 51871-62-6
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty