Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-4-pyridylalanine 95%

Supplier: Origen Biomedical, Inc

H52193-03 H52193-MD
AAH52193-03EA 209 USD InStock
AAH52193-03 AAH52193-06 AAH52193-MD
Fmoc-D-4-pyridylalanine 95%
Fmoc-D-4-pyridylalanine
Purity: 95% CAS No.: 205528-30-9 Molecular Formula: C23H20N2O4 Formula Weight: 388.42 MDL No.: MFCD00672567


Formula: C₂₃H₂₀N₂O₄
MW: 388.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672567
CAS Number: 205528-30-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty