Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one 96%

Supplier: Origen Biomedical, Inc

H52317-03 H52317-MD
AAH52317-03EA 503 USD InStock
AAH52317-03 AAH52317-MD
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one 96%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one
Purity: 96% CAS No.: 376584-62-2 Molecular Formula: C14H18BNO3 Formula Weight: 259.11 MDL No.: MFCD11040393


Formula: C₁₄H₁₈BNO₃
Storage Temperature: Ambient
MDL Number: MFCD11040393
CAS Number: 376584-62-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty